Assay Details
Gene Symbol: VPS37A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046161.2 | NP_001039626.1 | 3 - 4 | 592 | 148 | |
GenBank mRNA | BC114056.1 | - | 3 - 4 | 593 | 148 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
