Assay Details
Gene Symbol: PHB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046198.1 | NP_001039663.1 | 3 - 4 | 357 | 72 | |
GenBank mRNA | BC113241.1 | - | 3 - 4 | 357 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
