Assay Details
Gene Symbol: ABR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046204.2 | NP_001039669.2 | 21 - 22 | 2625 | 86 | |
GenBank mRNA | BC114145.1 | - | 7 - 8 | 869 | 86 | |
BC148971.1 | - | 21 - 22 | 2489 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
