Assay Details
Gene Symbol: TENT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046361.1 | NP_001039826.1 | 4 - 5 | 644 | 84 | |
GenBank mRNA | BC113319.1 | - | 4 - 5 | 644 | 84 | |
BT025394.1 | - | 3 - 4 | 452 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
