Assay Details
Gene Symbol: CLPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046414.2 | NP_001039879.1 | 3 - 4 | 400 | 114 | |
GenBank mRNA | BC112880.1 | - | 3 - 4 | 401 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
