Assay Details
Gene Symbol: CYLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046417.1 | NP_001039882.1 | 15 - 16 | 2637 | 98 | |
GenBank mRNA | BC114710.1 | - | 15 - 16 | 2637 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
