Assay Details
Gene Symbol: CDC42EP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046471.1 | NP_001039936.1 | 1 | 2547 | 150 | |
GenBank mRNA | BC114767.1 | - | 1 | 2547 | 150 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
