Assay Details
Gene Symbol: ADCYAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046555.1 | NP_001040020.1 | 2 - 3 | 239 | 139 | |
GenBank mRNA | AY924308.1 | - | 2 - 3 | 239 | 139 | |
BC123523.1 | - | 3 - 4 | 875 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
