Assay Details
Gene Symbol: COTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046593.1 | NP_001040058.1 | 3 - 4 | 491 | 79 | |
GenBank mRNA | BC113301.1 | - | 3 - 4 | 491 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
