Assay Details
Gene Symbol: DNASE2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075127.2 | NP_001068595.1 | 4 - 5 | 656 | 99 | |
GenBank mRNA | AF047017.1 | - | 2 - 3 | 251 | 99 | |
BC118429.1 | - | 4 - 5 | 657 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
