Assay Details
Gene Symbol: MPDU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075179.1 | NP_001068647.1 | 1 - 2 | 105 | 82 | |
GenBank mRNA | BC118438.1 | - | 1 - 2 | 105 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
