Assay Details
Gene Symbol: NKIRAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075387.1 | NP_001068855.1 | 1 - 2 | 186 | 74 | |
GenBank mRNA | BT025402.1 | - | 1 - 2 | 186 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001075387.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
