Assay Details
Gene Symbol: NKIRAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075387.1 | NP_001068855.1 | 4 - 5 | 1142 | 86 | |
GenBank mRNA | BT025402.1 | - | 4 - 5 | 1142 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
