Assay Details
Gene Symbol: CDC42EP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075495.1 | NP_001068963.1 | 1 - 2 | 78 | 86 | |
GenBank mRNA | BC118148.1 | - | 1 - 2 | 78 | 86 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC118148.1, NM_001075495.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
