Assay Details
Gene Symbol: UNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075513.2 | NP_001068981.1 | 4 - 5 | 727 | 101 | |
GenBank mRNA | BC118447.1 | - | 4 - 5 | 728 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
