Assay Details
Gene Symbol: SHPK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075548.1 | NP_001069016.1 | 1 - 2 | 187 | 122 | |
GenBank mRNA | BT025410.1 | - | 1 - 2 | 187 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
