Assay Details
Gene Symbol: SLC7A6OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075567.1 | NP_001069035.1 | 2 - 3 | 487 | 88 | |
GenBank mRNA | BC116007.1 | - | 2 - 3 | 487 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
