Assay Details
Gene Symbol: TOE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075594.1 | NP_001069062.1 | 1 - 2 | 191 | 84 | |
GenBank mRNA | BC118261.1 | - | 1 - 2 | 191 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
