Assay Details
Gene Symbol: NSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075595.3 | NP_001069063.3 | 7 - 8 | 2186 | 101 | |
GenBank mRNA | BC118092.1 | - | 7 - 8 | 1809 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
