Assay Details
Gene Symbol: APOO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075639.1 | NP_001069107.1 | 8 - 9 | 643 | 137 | |
GenBank mRNA | BC118339.1 | - | 8 - 9 | 643 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
