Assay Details
Gene Symbol: NAA60 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075649.1 | NP_001069117.1 | 1 - 2 | 146 | 74 | |
GenBank mRNA | BC118298.1 | - | 1 - 2 | 146 | 74 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC118298.1, NM_001075649.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
