Assay Details
Gene Symbol: IDI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075659.1 | NP_001069127.1 | 4 - 5 | 778 | 85 | |
GenBank mRNA | BC116133.1 | - | 4 - 5 | 778 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
