Assay Details
Gene Symbol: SDR39U1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075812.2 | NP_001069280.1 | 5 - 6 | 541 | 84 | |
GenBank mRNA | BC118349.1 | - | 5 - 6 | 543 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
