Assay Details
Gene Symbol: SLC52A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075901.2 | NP_001069369.1 | 1 - 2 | 468 | 69 | |
GenBank mRNA | BC118403.1 | - | 1 - 2 | 469 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC118403.1, NM_001075901.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
