Assay Details
Gene Symbol: SLC52A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075901.2 | NP_001069369.1 | 4 - 5 | 1654 | 91 | |
GenBank mRNA | BC118403.1 | - | 4 - 5 | 1655 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
