Assay Details
Gene Symbol: MRPS31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076015.2 | NP_001069483.1 | 3 - 4 | 611 | 75 | |
GenBank mRNA | BC118356.1 | - | 3 - 4 | 612 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
