Assay Details
Gene Symbol: NCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076072.1 | NP_001069540.1 | 1 - 2 | 124 | 87 | |
GenBank mRNA | BC116109.1 | - | 1 - 2 | 124 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC116109.1, NM_001076072.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
