Assay Details
Gene Symbol: BCL11A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076121.2 | NP_001069589.1 | 2 - 3 | 755 | 121 | |
GenBank mRNA | BC118227.1 | - | 2 - 3 | 613 | 121 | |
LC016540.1 | - | 2 - 3 | 473 | 121 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
