Assay Details
Gene Symbol: SUPT7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076150.2 | NP_001069618.1 | 2 - 3 | 339 | 103 | |
GenBank mRNA | BC114175.1 | - | 2 - 3 | 340 | 103 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC114175.1, NM_001076150.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
