Assay Details
Gene Symbol: SUPT7L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076150.2 | NP_001069618.1 | 5 - 6 | 1518 | 90 | |
GenBank mRNA | BC114175.1 | - | 5 - 6 | 1519 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
