Assay Details
Gene Symbol: CFL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076154.2 | NP_001069622.1 | 3 - 4 | 408 | 126 | |
GenBank mRNA | BC118390.1 | - | 3 - 4 | 410 | 126 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
