Assay Details
Gene Symbol: ALOX5AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076293.2 | NP_001069761.1 | 2 - 3 | 205 | 71 | |
GenBank mRNA | BC118389.1 | - | 2 - 3 | 206 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
