Assay Details
Gene Symbol: ALOX5AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076293.2 | NP_001069761.1 | 4 - 5 | 354 | 81 | |
GenBank mRNA | BC118389.1 | - | 4 - 5 | 355 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
