Assay Details
Gene Symbol: TIGAR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076370.1 | NP_001069838.1 | 3 - 4 | 233 | 132 | |
GenBank mRNA | BC116101.1 | - | 3 - 4 | 233 | 132 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
