Assay Details
Gene Symbol: EFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076432.1 | NP_001069900.1 | 3 - 4 | 731 | 128 | |
GenBank mRNA | BC118089.1 | - | 3 - 4 | 731 | 128 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
