Assay Details
Gene Symbol: POLR2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076458.2 | NP_001069926.1 | 1 - 2 | 98 | 95 | |
GenBank mRNA | BC116155.1 | - | 1 - 2 | 99 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
