Assay Details
Gene Symbol: UBE2S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076472.2 | NP_001069940.1 | 1 - 2 | 239 | 141 | |
GenBank mRNA | BC114838.1 | - | 1 - 2 | 240 | 141 | |
BC148138.1 | - | 1 - 2 | 202 | 141 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC114838.1, BC148138.1, NM_001076472.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
