Assay Details
Gene Symbol: DOC2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076499.2 | NP_001069967.1 | 6 - 7 | 800 | 65 | |
GenBank mRNA | BC118129.1 | - | 6 - 7 | 801 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
