Assay Details
Gene Symbol: MS4A14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076541.1 | NP_001070009.1 | 4 - 5 | 560 | 83 | |
GenBank mRNA | BC114037.1 | - | 4 - 5 | 560 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
