Assay Details
Gene Symbol: DHFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077883.1 | NP_001071351.1 | 3 - 4 | 315 | 109 | |
GenBank mRNA | BC114162.1 | - | 3 - 4 | 315 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
