Assay Details
Gene Symbol: HSP90AB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079637.1 | NP_001073105.1 | 2 - 3 | 232 | 119 | |
GenBank mRNA | AB072369.1 | - | 2 - 3 | 227 | 119 | |
BC102944.1 | - | 2 - 3 | 246 | 119 | ||
BT025368.1 | - | 2 - 3 | 232 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
