Assay Details
Gene Symbol: HSP90AB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079637.1 | NP_001073105.1 | 4 - 5 | 603 | 78 | |
GenBank mRNA | AB072369.1 | - | 4 - 5 | 598 | 78 | |
BC102944.1 | - | 4 - 5 | 617 | 78 | ||
BT025368.1 | - | 4 - 5 | 603 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
