Assay Details
Gene Symbol: LRPAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080225.1 | NP_001073694.1 | 1 - 2 | 226 | 65 | |
GenBank mRNA | BC118191.1 | - | 1 - 2 | 226 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
