Assay Details
Gene Symbol: FOXRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034371.3 | NP_001029543.2 | 4 - 5 | 637 | 59 | |
GenBank mRNA | BC120036.1 | - | 4 - 5 | 638 | 59 | |
BT020666.1 | - | 4 - 5 | 583 | 59 | ||
BT020724.1 | - | 4 - 5 | 584 | 59 | ||
BT021902.1 | - | 4 - 5 | 586 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
