Assay Details
Gene Symbol: CNIH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034739.2 | NP_001029911.1 | 4 - 5 | 413 | 100 | |
GenBank mRNA | BC119865.1 | - | 4 - 5 | 413 | 100 | |
BT021188.1 | - | 4 - 5 | 413 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
