Assay Details
Gene Symbol: FEZF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038198.2 | NP_001033287.1 | 2 - 3 | 162 | 88 | |
GenBank mRNA | BC123390.1 | - | 2 - 3 | 162 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC123390.1, NM_001038198.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
