Assay Details
Gene Symbol: ETAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075128.1 | NP_001068596.1 | 3 - 4 | 548 | 93 | |
GenBank mRNA | BC123739.1 | - | 3 - 4 | 548 | 93 | |
Y08459.1 | - | 527 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
