Assay Details
Gene Symbol: ETAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075128.1 | NP_001068596.1 | 5 - 6 | 2693 | 114 | |
GenBank mRNA | BC123739.1 | - | 5 - 6 | 2693 | 114 | |
Y08459.1 | - | 2672 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
