Assay Details
Gene Symbol: GCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075134.1 | NP_001068602.1 | 3 - 4 | 433 | 72 | |
GenBank mRNA | AF195767.1 | - | 3 - 4 | 354 | 72 | |
BC119870.1 | - | 3 - 4 | 433 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
