Assay Details
Gene Symbol: GCAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075134.1 | NP_001068602.1 | 5 - 6 | 734 | 56 | |
GenBank mRNA | AF195767.1 | - | 5 - 6 | 655 | 56 | |
BC119870.1 | - | 5 - 6 | 734 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
